Assay Details
Gene Symbol: LOC102174197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045398.1 | XP_017900887.1 | 1669 | 66 | ||
GenBank mRNA | JO425056.1 | - | 1973 | 66 |
Gene Symbol: LOC108634427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO402213.2 | - | 1913 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
