Assay Details
Gene Symbol: LOC102183271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO631943.2 | - | 1786 | 103 |
Gene Symbol: APBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052054.1 | XP_017907543.1 | 5 - 6 | 1738 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
