Assay Details
Gene Symbol: SGMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681329.3 | XP_005681386.1 | 3 - 4 | 1090 | 78 | |
XM_018049235.1 | XP_017904724.1 | 3 - 4 | 1148 | 78 | ||
GenBank mRNA | JT622483.1 | - | 4 - 5 | 779 | 78 | |
JT662938.1 | - | 4 - 5 | 803 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
