Assay Details
Gene Symbol: ARHGAP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058758.1 | XP_017914247.1 | 6 - 7 | 2861 | 75 | |
XM_018058759.1 | XP_017914248.1 | 8 - 9 | 2868 | 75 | ||
XM_018058760.1 | XP_017914249.1 | 7 - 8 | 2775 | 75 | ||
XM_018058761.1 | XP_017914250.1 | 8 - 9 | 2655 | 75 | ||
GenBank mRNA | KA322698.1 | - | 4 - 5 | 1714 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
