Assay Details
Gene Symbol: ZNF608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965358.2 | XP_013820812.2 | 2 - 3 | 1884 | 62 | |
XM_018050692.1 | XP_017906181.1 | 3 - 4 | 2723 | 62 | ||
XM_018050693.1 | XP_017906182.1 | 2 - 3 | 1912 | 62 | ||
XM_018050695.1 | XP_017906184.1 | 2 - 3 | 503 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
