Assay Details
| Gene Symbol: RRAGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053149.1 | XP_017908638.1 | 2 - 3 | 668 | 62 | |
| GenBank mRNA | JO455519.2 | - | 1 - 2 | 179 | 62 | |
More Information
Set Membership: |
|

RRAGD
Genomic Map