Assay Details
Gene Symbol: GPR180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013968320.2 | XP_013823774.2 | 4 - 5 | 1025 | 85 | |
GenBank mRNA | JO602347.2 | - | 4 - 5 | 701 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
