Assay Details
| Gene Symbol: FOXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053854.1 | XP_017909343.1 | 4 - 5 | 893 | 78 | |
| XM_018053855.1 | XP_017909344.1 | 3 - 4 | 845 | 78 | ||
| XM_018053856.1 | XP_017909345.1 | 4 - 5 | 892 | 78 | ||
| XM_018053857.1 | XP_017909346.1 | 3 - 4 | 845 | 78 | ||
| GenBank mRNA | JO408668.2 | - | 4 - 5 | 830 | 78 | |
| JO595352.2 | - | 4 - 5 | 972 | 78 | ||
More Information
Set Membership: |
|

FOXN3
Genomic Map