Assay Details
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693262.3 | XP_005693319.1 | 3 - 4 | 698 | 80 | |
XM_005693263.3 | XP_005693320.1 | 2 - 3 | 611 | 80 | ||
XM_005693264.3 | XP_005693321.1 | 2 - 3 | 448 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
