Assay Details
Gene Symbol: TBXA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682579.3 | XP_005682636.2 | 2 - 3 | 1209 | 106 | |
XM_018050831.1 | XP_017906320.1 | 1 - 2 | 786 | 106 | ||
GenBank mRNA | JO397474.2 | - | 1 - 2 | 1190 | 106 | |
KA323030.1 | - | 1 - 2 | 1210 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
