Assay Details
Gene Symbol: UPK3BL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697803.3 | XP_005697860.3 | 4 - 5 | 921 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UPK3BL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697803.3 | XP_005697860.3 | 4 - 5 | 921 | 99 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |