Assay Details
| Gene Symbol: KLF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058329.1 | XP_017913818.1 | 3 - 4 | 1590 | 71 | |
| GenBank mRNA | KU643198.1 | - | 3 - 4 | 1190 | 71 | |
More Information
Set Membership: |
|

KLF10
Genomic Map