Assay Details
| Gene Symbol: OAZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290529.1 | NP_001277458.1 | 2 - 3 | 335 | 60 | |
| NM_001301483.1 | NP_001288412.1 | 2 - 3 | 367 | 60 | ||
More Information
Set Membership: |
|

OAZ3
Genomic Map