Assay Details
Gene Symbol: CNNM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041157.1 | XP_017896646.1 | 4 - 5 | 2252 | 74 | |
XM_018041158.1 | XP_017896647.1 | 4 - 5 | 2252 | 74 | ||
GenBank mRNA | JI861758.1 | - | 4 - 5 | 1524 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
