Assay Details
| Gene Symbol: LZTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057417.1 | XP_017912906.1 | 3 - 4 | 808 | 95 | |
| XM_018057418.1 | XP_017912907.1 | 3 - 4 | 4731 | 95 | ||
| XM_018057420.1 | XP_017912909.1 | 3 - 4 | 767 | 95 | ||
| XM_018057421.1 | XP_017912910.1 | 3 - 4 | 770 | 95 | ||
| XM_018057422.1 | XP_017912911.1 | 2 - 3 | 588 | 95 | ||
More Information
Set Membership: |
|

LZTS3
Genomic Map