Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677729.3 | XP_005677786.2 | 2 - 3 | 365 | 81 | |
XM_005677730.3 | XP_005677787.2 | 2 - 3 | 383 | 81 | ||
GenBank mRNA | JO411586.2 | - | 2 - 3 | 1847 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
