Assay Details
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044095.1 | XP_017899584.1 | 4 - 5 | 559 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
