Assay Details
| Gene Symbol: POPDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684614.3 | XP_005684671.1 | 3 - 4 | 918 | 79 | |
| XM_018053119.1 | XP_017908608.1 | 3 - 4 | 1050 | 79 | ||
| XM_018053120.1 | XP_017908609.1 | 4 - 5 | 1112 | 79 | ||
More Information
Set Membership: |
|

POPDC3
Genomic Map