Assay Details
Gene Symbol: NR3C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691196.3 | XP_005691253.2 | 5 - 6 | 2882 | 68 | |
XM_013970502.2 | XP_013825956.2 | 5 - 6 | 2923 | 68 | ||
XM_018061588.1 | XP_017917077.1 | 5 - 6 | 2419 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
