Assay Details
| Gene Symbol: LOC108635581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045824.1 | XP_017901313.1 | 17 - 18 | 2482 | 76 | |
| XM_018045830.1 | XP_017901319.1 | 17 - 18 | 2480 | 76 | ||
| XM_018045834.1 | XP_017901323.1 | 18 - 19 | 2583 | 76 | ||
More Information
Set Membership: |
|

LOC108635581
Genomic Map