Assay Details
| Gene Symbol: ZNRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691881.3 | XP_005691938.3 | 2 - 3 | 820 | 60 | |
| XM_018061707.1 | XP_017917196.1 | 2 - 3 | 820 | 60 | ||
| XM_018061708.1 | XP_017917197.1 | 2 - 3 | 820 | 60 | ||
| XM_018061709.1 | XP_017917198.1 | 2 - 3 | 820 | 60 | ||
| GenBank mRNA | JO364249.2 | - | 2 - 3 | 1042 | 60 | |
| KA325250.1 | - | 2 - 3 | 713 | 60 | ||
More Information
Set Membership: |
|

ZNRF1
Genomic Map