Assay Details
| Gene Symbol: NIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692185.3 | XP_005692242.1 | 1 - 2 | 231 | 66 | |
| XM_005692186.3 | XP_005692243.1 | 1 - 2 | 242 | 66 | ||
| GenBank mRNA | JO585874.2 | - | 1 - 2 | 155 | 66 | |
| JT702505.1 | - | 4 - 5 | 1638 | 66 | ||
More Information
Set Membership: |
|

NIP7
Genomic Map