Assay Details
Gene Symbol: NPTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063634.1 | XP_017919123.1 | 3 - 4 | 1042 | 67 | |
GenBank mRNA | JO643449.2 | - | 3 - 4 | 1002 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
