Assay Details
| Gene Symbol: ZNF384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048563.1 | XP_017904052.1 | 12 - 13 | 1761 | 71 | |
| XM_018048564.1 | XP_017904053.1 | 11 - 12 | 1704 | 71 | ||
| XM_018048565.1 | XP_017904054.1 | 11 - 12 | 2065 | 71 | ||
| XM_018048566.1 | XP_017904055.1 | 11 - 12 | 1523 | 71 | ||
| XM_018048567.1 | XP_017904056.1 | 10 - 11 | 1700 | 71 | ||
| XM_018048568.1 | XP_017904057.1 | 10 - 11 | 1653 | 71 | ||
| XM_018048569.1 | XP_017904058.1 | 10 - 11 | 1621 | 71 | ||
| XM_018048570.1 | XP_017904059.1 | 10 - 11 | 1565 | 71 | ||
| XM_018048571.1 | XP_017904060.1 | 9 - 10 | 1470 | 71 | ||
| XM_018048572.1 | XP_017904061.1 | 8 - 9 | 1421 | 71 | ||
More Information
Set Membership: |
|

ZNF384
Genomic Map