Assay Details
Gene Symbol: FOSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055489.1 | XP_017910978.1 | 2 - 3 | 1176 | 84 | |
GenBank mRNA | JO328752.2 | - | 2 - 3 | 407 | 84 | |
KA314323.1 | - | 2 - 3 | 1205 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
