Assay Details
Gene Symbol: LRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058336.1 | XP_017913825.1 | 4 - 5 | 599 | 61 | |
XM_018058337.1 | XP_017913826.1 | 3 - 4 | 542 | 61 | ||
GenBank mRNA | JT683177.1 | - | 3 - 4 | 397 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
