Assay Details
Gene Symbol: AURKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693002.3 | XP_005693059.1 | 3 - 4 | 483 | 62 | |
XM_005693003.3 | XP_005693060.1 | 3 - 4 | 480 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
