Assay Details
| Gene Symbol: SSNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056262.1 | XP_017911751.1 | 2 - 3 | 310 | 85 | |
| GenBank mRNA | JO645520.2 | - | 1 - 2 | 524 | 85 | |
| JO645521.2 | - | 1 - 2 | 524 | 85 | ||
More Information
Set Membership: |
|

SSNA1
Genomic Map