Assay Details
Gene Symbol: GORASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018067074.1 | XP_017922563.1 | 2 - 3 | 240 | 67 | |
XM_018067075.1 | XP_017922564.1 | 5 - 6 | 566 | 67 | ||
XM_018067076.1 | XP_017922565.1 | 4 - 5 | 652 | 67 | ||
XM_018067077.1 | XP_017922566.1 | 2 - 3 | 711 | 67 | ||
XM_018067078.1 | XP_017922567.1 | 4 - 5 | 458 | 67 | ||
GenBank mRNA | JO633126.2 | - | 2 - 3 | 294 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018067078.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
