Assay Details
| Gene Symbol: IL12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285700.1 | NP_001272629.1 | 5 - 6 | 853 | 61 | |
| GenBank mRNA | AF007576.1 | - | 5 - 6 | 853 | 61 | |
| AY603406.1 | - | 5 - 6 | 853 | 61 | ||
More Information
Set Membership: |
|

IL12B
Genomic Map