Assay Details
Gene Symbol: SLC2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285741.1 | NP_001272670.1 | 6 - 7 | 964 | 66 | |
GenBank mRNA | GU591154.1 | - | 6 - 7 | 964 | 66 | |
JT689633.1 | - | 8 - 9 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
