Assay Details
Gene Symbol: SDF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285643.1 | NP_001272572.1 | 1 - 2 | 302 | 58 | |
XM_018059911.1 | XP_017915400.1 | 2 - 3 | 463 | 58 | ||
XM_018059912.1 | XP_017915401.1 | 2 - 3 | 471 | 58 | ||
XM_018059913.1 | XP_017915402.1 | 2 - 3 | 439 | 58 | ||
GenBank mRNA | EF589950.1 | - | 1 - 2 | 302 | 58 | |
JO355186.2 | - | 2 - 3 | 487 | 58 | ||
JO500258.2 | - | 2 - 3 | 527 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
