Assay Details
Gene Symbol: HSPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287571.1 | NP_001274500.1 | 5 - 6 | 1002 | 64 | |
XM_005687138.3 | XP_005687195.1 | 6 - 7 | 1198 | 64 | ||
GenBank mRNA | KC292214.1 | - | 5 - 6 | 1002 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
