Assay Details
Gene Symbol: BTG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289948.1 | NP_001276877.1 | 2 - 3 | 228 | 85 | |
GenBank mRNA | JO352263.2 | - | 3 - 4 | 701 | 85 | |
JO388690.2 | - | 3 - 4 | 715 | 85 | ||
JO446296.2 | - | 1 - 2 | 138 | 85 | ||
JO526704.3 | - | 2 - 3 | 259 | 85 | ||
JO548702.2 | - | 3 - 4 | 647 | 85 | ||
JO565982.2 | - | 1 - 2 | 168 | 85 | ||
JO575245.2 | - | 1 - 2 | 168 | 85 | ||
JO586623.2 | - | 3 - 4 | 678 | 85 | ||
JT706680.1 | - | 2 - 3 | 322 | 85 | ||
JT716838.1 | - | 2 - 3 | 226 | 85 | ||
JT738521.1 | - | 1 - 2 | 168 | 85 | ||
KA327599.1 | - | 3 - 4 | 730 | 85 | ||
KC537060.1 | - | 2 - 3 | 228 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
