Assay Details
| Gene Symbol: ELOVL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314257.1 | NP_001301186.1 | 2 - 3 | 209 | 74 | |
| XM_005681307.3 | XP_005681364.1 | 3 - 4 | 349 | 74 | ||
| XM_005681308.3 | XP_005681365.1 | 3 - 4 | 420 | 74 | ||
| GenBank mRNA | KA337802.1 | - | 2 - 3 | 3039 | 74 | |
| KF667508.1 | - | 2 - 3 | 209 | 74 | ||
More Information
Set Membership: |
|

ELOVL6
Genomic Map