Assay Details
| Gene Symbol: EPHX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285557.1 | NP_001272486.1 | 1 - 2 | 189 | 73 | |
| XM_013970074.2 | XP_013825528.2 | 2 - 3 | 327 | 73 | ||
| XM_013970075.2 | XP_013825529.2 | 2 - 3 | 208 | 73 | ||
| XM_018059920.1 | XP_017915409.1 | 2 - 3 | 317 | 73 | ||
| GenBank mRNA | HQ326238.1 | - | 1 - 2 | 192 | 73 | |
| JO419527.2 | - | 2 - 3 | 495 | 73 | ||
More Information
Set Membership: |
|

EPHX1
Genomic Map