Assay Details
Gene Symbol: TLR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285574.1 | NP_001272503.1 | 2 - 3 | 375 | 98 | |
GenBank mRNA | FJ659851.1 | - | 1 - 2 | 61 | 98 | |
JF825527.1 | - | 2 - 3 | 375 | 98 | ||
JO519293.2 | - | 2 - 3 | 530 | 98 | ||
JQ342090.1 | - | 1 - 2 | 60 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
