Assay Details
| Gene Symbol: TLR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285699.1 | NP_001272628.1 | 1 | 2517 | 95 | |
| XM_013970071.2 | XP_013825525.2 | 2 | 3151 | 95 | ||
| XM_018059914.1 | XP_017915403.1 | 3 | 2766 | 95 | ||
| XM_018059915.1 | XP_017915404.1 | 4 | 3041 | 95 | ||
| XM_018059916.1 | XP_017915405.1 | 2 | 2597 | 95 | ||
| GenBank mRNA | FJ659852.1 | - | 1 | 2517 | 95 | |
More Information
Set Membership: |
|

TLR5
Genomic Map