Assay Details
Gene Symbol: TNFSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285702.1 | NP_001272631.1 | 3 - 4 | 488 | 64 | |
XM_018056283.1 | XP_017911772.1 | 3 - 4 | 687 | 64 | ||
XM_018056284.1 | XP_017911773.1 | 3 - 4 | 690 | 64 | ||
GenBank mRNA | FJ793192.1 | - | 3 - 4 | 488 | 64 | |
JT681577.1 | - | 586 | 64 | |||
JT700368.1 | - | 3 - 4 | 728 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
