Assay Details
Gene Symbol: SDHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285731.1 | NP_001272660.1 | 3 - 4 | 343 | 72 | |
GenBank mRNA | GU338978.1 | - | 3 - 4 | 346 | 72 | |
KA307331.1 | - | 1 - 2 | 760 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
