Assay Details
Gene Symbol: PROLACTIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285547.1 | NP_001272476.1 | 252 | 98 | ||
GenBank mRNA | JF938531.1 | - | 255 | 98 | ||
X76048.1 | - | 254 | 98 | |||
X76049.1 | - | 254 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
