Assay Details
| Gene Symbol: GHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285648.1 | NP_001272577.1 | 2 - 3 | 361 | 70 | |
| XM_013972754.2 | XP_013828208.1 | 2 - 3 | 98 | 70 | ||
| XM_013972755.2 | XP_013828209.1 | 1 - 2 | 89 | 70 | ||
| XM_013972756.2 | XP_013828210.1 | 2 - 3 | 319 | 70 | ||
| XM_018065463.1 | XP_017920952.1 | 2 - 3 | 197 | 70 | ||
| XM_018065464.1 | XP_017920953.1 | 2 - 3 | 140 | 70 | ||
| GenBank mRNA | EF559245.1 | - | 2 - 3 | 361 | 70 | |
More Information
Set Membership: |
|

GHR
Genomic Map