Assay Details
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286965.1 | NP_001273894.1 | 4 - 5 | 548 | 74 | |
GenBank mRNA | EU244428.1 | - | 4 - 5 | 282 | 74 | |
JO377265.2 | - | 4 - 5 | 548 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
