Assay Details
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314194.1 | NP_001301123.1 | 5 - 6 | 563 | 84 | |
XM_018039493.1 | XP_017894982.1 | 5 - 6 | 699 | 84 | ||
GenBank mRNA | JO613209.2 | - | 4 - 5 | 542 | 84 | |
KP997221.1 | - | 5 - 6 | 563 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
