Assay Details
| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285539.1 | NP_001272468.1 | 1 - 2 | 139 | 58 | |
| XM_018054687.1 | XP_017910176.1 | 1 - 2 | 315 | 58 | ||
| XM_018054688.1 | XP_017910177.1 | 1 - 2 | 315 | 58 | ||
| XM_018054689.1 | XP_017910178.1 | 1 - 2 | 315 | 58 | ||
| GenBank mRNA | EF564255.1 | - | 1 - 2 | 129 | 58 | |
| JO517063.2 | - | 6 - 7 | 552 | 58 | ||
| X58471.1 | - | 1 - 2 | 111 | 58 | ||
| Z19569.1 | - | 1 - 2 | 139 | 58 | ||
More Information
Set Membership: |
|

PAEP
Genomic Map