Assay Details
| Gene Symbol: GNRHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285612.1 | NP_001272541.1 | 2 - 3 | 749 | 77 | |
| XM_005681659.2 | XP_005681716.2 | 2 - 3 | 614 | 77 | ||
| GenBank mRNA | EF150356.1 | - | 2 - 3 | 749 | 77 | |
More Information
Set Membership: |
|

GNRHR
Genomic Map