Assay Details
| Gene Symbol: ANGPTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285709.1 | NP_001272638.1 | 6 - 7 | 1252 | 87 | |
| GenBank mRNA | GQ293360.1 | - | 6 - 7 | 1252 | 87 | |
| JO419837.2 | - | 1 - 2 | 416 | 87 | ||
More Information
Set Membership: |
|

ANGPTL3
Genomic Map