Assay Details
Gene Symbol: KRT26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285714.1 | NP_001272643.1 | 1 - 2 | 464 | 90 | |
XM_013971961.2 | XP_013827415.2 | 1 - 2 | 499 | 90 | ||
GenBank mRNA | GU084153.1 | - | 1 - 2 | 464 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
