Assay Details
| Gene Symbol: PEG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291299.1 | NP_001278228.1 | 1 - 2 | 208 | 78 | |
| NM_001291300.1 | NP_001278229.1 | 1 - 2 | 208 | 78 | ||
| GenBank mRNA | JO450726.2 | - | 1 - 2 | 240 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001291299.1, NM_001291300.1 transcripts.
Set Membership: |
|

PEG10
Genomic Map