Assay Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285544.1 | NP_001272473.1 | 4 - 5 | 256 | 108 | |
| GenBank mRNA | AY524275.1 | - | 1 - 2 | 118 | 108 | |
| AY605263.1 | - | 4 - 5 | 256 | 108 | ||
| JO586553.2 | - | 4 - 5 | 374 | 108 | ||
| KA347557.1 | - | 2 - 3 | 376 | 108 | ||
More Information
Set Membership: |
|

IL18
Genomic Map